2-Methyl-1-(4-propoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

331141-1G 331141-500MG 331141-5G 331141-25G
77469-528EA 260.59 USD
77469-528 77482-204 77492-608 77502-462
2-Methyl-1-(4-propoxyphenyl)propan-1-one ≥95%
2-Methyl-1-(4-propoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
CAS Number: 64436-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR