2-(4-propoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

331134-1G 331134-500MG 331134-5G 331134-25G
77469-524EA 415.09 USD
77469-524 77482-200 77492-604 77502-458
2-(4-propoxyphenyl)propan-2-ol ≥95%
2-(4-propoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Storage Temperature: Ambient
CAS Number: 1342021-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR