phenyl(4-propoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

331132-1G 331132-500MG 331132-5G 331132-25G
77469-520EA 239.99 USD
77469-520 77482-196 77492-600 77502-454
phenyl(4-propoxyphenyl)methanone ≥95%
phenyl(4-propoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₆H₁₆O₂
Storage Temperature: Ambient
CAS Number: 65565-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR