1-(4-(cyclopentyloxy)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

331127-1G 331127-500MG 331127-5G
77469-516EA 676.71 USD
77469-516 77482-192 77492-596
1-(4-(cyclopentyloxy)phenyl)prop-2-en-1-ol ≥95%
1-(4-(cyclopentyloxy)phenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₄H₁₈O₂
MW: 218.292 g/mol
Storage Temperature: Ambient
CAS Number: 1508758-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR