(4-(cyclopentyloxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

331116-1G 331116-500MG 331116-5G
77469-512EA 481.01 USD
77469-512 77482-188 77492-592
(4-(cyclopentyloxy)phenyl)(methyl)sulfane ≥95%
(4-(cyclopentyloxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₆OS
MW: 208.32 g/mol
Storage Temperature: Ambient
CAS Number: 3002444-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR