4-(Cyclopentyloxy)-benzoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(cyclopentyloxy)benzoic acid

331110-1G 331110-500MG 331110-5G 331110-25G
77469-508EA 103 USD
77469-508 77482-184 77492-588 77502-446
4-(Cyclopentyloxy)-benzoic acid ≥95%
4-(Cyclopentyloxy)-benzoic acid

Precaución: For research use only

Formula: C₁₂H₁₄O₃
MW: 206.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03233617
CAS Number: 30762-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR