4-Cyclohexyloxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Cyclohexyloxy)benzoic acid

331091-1G 331091-500MG 331091-5G 331091-25G
77469-504EA 167.89 USD
77469-504 77482-180 77492-584 77502-442
4-Cyclohexyloxybenzoic acid ≥95%
4-Cyclohexyloxybenzoic acid

Precaución: For research use only

Formula: C₁₃H₁₆O₃
MW: 220.27 g/mol
Storage Temperature: Ambient
CAS Number: 139-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR