1-(3-Ethoxy-5-(trifluoromethyl)phenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

331065-1G 331065-500MG 331065-5G 331065-25G
77469-500EA 415.09 USD
77469-500 77482-176 77492-580 77502-438
1-(3-Ethoxy-5-(trifluoromethyl)phenyl)-2-ethylbutan-1-ol ≥95%
1-(3-Ethoxy-5-(trifluoromethyl)phenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₁F₃O₂
MW: 290.321 g/mol
Storage Temperature: Ambient
CAS Number: 3002439-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR