1-(3-Ethoxy-5-(trifluoromethyl)phenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

331061-1G 331061-500MG 331061-5G 331061-25G
77469-496EA 415.09 USD
77469-496 77482-172 77492-576 77502-434
1-(3-Ethoxy-5-(trifluoromethyl)phenyl)cycloheptanol ≥95%
1-(3-Ethoxy-5-(trifluoromethyl)phenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₆H₂₁F₃O₂
MW: 302.332 g/mol
Storage Temperature: Ambient
CAS Number: 3002454-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR