2-(3-Ethoxy-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

331056-1G 331056-500MG 331056-5G 331056-25G
77469-492EA 415.09 USD
77469-492 77482-168 77492-572 77502-430
2-(3-Ethoxy-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(3-Ethoxy-5-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₅F₃O₂
MW: 248.242 g/mol
Storage Temperature: Ambient
CAS Number: 3002511-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR