2-Fluoro-4-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

33100-1G 33100-500MG 33100-5G
77469-488EA 830.18 USD
77469-488 77482-164 77492-568
2-Fluoro-4-methylpyridin-3-ol ≥97%
2-Fluoro-4-methylpyridin-3-ol

Precaución: For research use only

Formula: C₆H₆FNO
MW: 127.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611069
CAS Number: 1092561-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR