1-((3-bromobenzyl)oxy)-2-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

330729-1G 330729-500MG 330729-5G
77469-480EA 576.8 USD
77469-480 77482-156 77492-560
1-((3-bromobenzyl)oxy)-2-isopropylbenzene ≥95%
1-((3-bromobenzyl)oxy)-2-isopropylbenzene

Precaución: For research use only

Formula: C₁₆H₁₇BrO
MW: 305.21 g/mol
Storage Temperature: Ambient
CAS Number: 1275453-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR