1-(3-Bromo-6-chloro-2-fluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

330708-1G 330708-500MG 330708-5G 330708-25G
77469-476EA 415.09 USD
77469-476 77482-152 77492-556 77502-418
1-(3-Bromo-6-chloro-2-fluorophenyl)-2-methylpropan-1-ol ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrClFO
MW: 281.5491 g/mol
Storage Temperature: Ambient
CAS Number: 3002480-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR