1-(2-Methoxy-4,5-dimethylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

329715-1G 329715-500MG 329715-5G
77469-472EA 646.84 USD
77469-472 77482-148 77492-552
1-(2-Methoxy-4,5-dimethylphenyl)-1-propanone ≥95%
1-(2-Methoxy-4,5-dimethylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MW: 192.25 g/mol
Storage Temperature: Ambient
CAS Number: 36871-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR