6-Bromo-2,3-dichloroanisole ≥97%

Supplier: AOBChem USA

Warning

32929-1G 32929-500MG 32929-5G
77469-468EA 455.26 USD
77469-468 77482-144 77492-548
6-Bromo-2,3-dichloroanisole ≥97%
6-Bromo-2,3-dichloroanisole

Precaución: For research use only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD22414759
CAS Number: 174913-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR