(2-(Benzyloxy)-5-methylphenyl)(3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

328838-1G 328838-500MG 328838-5G 328838-25G
77469-464EA 415.09 USD
77469-464 77482-140 77492-544 77502-412
(2-(Benzyloxy)-5-methylphenyl)(3-methoxyphenyl)methanol ≥95%
(2-(Benzyloxy)-5-methylphenyl)(3-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₂₂H₂₂O₃
MW: 334.408 g/mol
Storage Temperature: Ambient
CAS Number: 3002480-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR