Cyclohexyl(2-ethoxy-5-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

328631-1G 328631-500MG 328631-5G 328631-25G
77469-460EA 415.09 USD
77469-460 77482-136 77492-540 77502-408
Cyclohexyl(2-ethoxy-5-methylphenyl)methanone ≥95%
Cyclohexyl(2-ethoxy-5-methylphenyl)methanone

Precaución: For research use only

Formula: C₁₆H₂₂O₂
Storage Temperature: Ambient
CAS Number: 1274750-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR