1-(2-Ethoxy-5-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

328629-1G 328629-500MG 328629-5G 328629-25G
77469-456EA 341.96 USD
77469-456 77482-132 77492-536 77502-404
1-(2-Ethoxy-5-methylphenyl)propan-1-one ≥95%
1-(2-Ethoxy-5-methylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MW: 192.2542 g/mol
Storage Temperature: Ambient
CAS Number: 1071378-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR