(2-Ethoxy-5-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

328627-1G 328627-500MG 328627-5G 328627-25G
77469-452EA 415.09 USD
77469-452 77482-128 77492-532 77502-400
(2-Ethoxy-5-methylphenyl)(phenyl)methanol ≥95%
(2-Ethoxy-5-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
CAS Number: 1272833-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR