(2-Ethoxy-5-methylphenyl)(3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

328620-1G 328620-500MG 328620-5G 328620-25G
77469-444EA 415.09 USD
77469-444 77482-120 77492-524 77502-392
(2-Ethoxy-5-methylphenyl)(3-methoxyphenyl)methanol ≥95%
(2-Ethoxy-5-methylphenyl)(3-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₇H₂₀O₃
Storage Temperature: Ambient
CAS Number: 1457119-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR