1-(4-Methoxyphenyl)-3-methyl-1-pentyn-3-ol ≥95%

Supplier: AOBChem USA

Warning

328168-1G 328168-500MG 328168-5G 328168-25G
77469-440EA 260.59 USD
77469-440 77482-116 77492-520 77502-388
1-(4-Methoxyphenyl)-3-methyl-1-pentyn-3-ol ≥95%
1-(4-Methoxyphenyl)-3-methyl-1-pentyn-3-ol

Precaución: For research use only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Storage Temperature: Ambient
CAS Number: 412013-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR