1-(4-Methoxyphenyl)-4-methyl-1-pentyn-3-ol ≥95%

Supplier: AOBChem USA

Warning

328150-1G 328150-500MG 328150-5G
77469-436EA 515 USD
77469-436 77482-112 77492-516
1-(4-Methoxyphenyl)-4-methyl-1-pentyn-3-ol ≥95%
1-(4-Methoxyphenyl)-4-methyl-1-pentyn-3-ol

Precaución: For research use only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Storage Temperature: Ambient
CAS Number: 88525-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR