4-(4-Methoxyphenyl)-3-butyn-2-one ≥95%

Supplier: AOBChem USA

Warning

328145-1G 328145-500MG 328145-5G 328145-25G
77469-432EA 364.62 USD
77469-432 77482-108 77492-512 77502-386
4-(4-Methoxyphenyl)-3-butyn-2-one ≥95%
4-(4-Methoxyphenyl)-3-butyn-2-one

Precaución: For research use only

Formula: C₁₁H₁₀O₂
Storage Temperature: Ambient
CAS Number: 81532-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR