1-(4'-Methyl[1,1'-biphenyl]-4-yl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

327707-1G 327707-500MG 327707-5G 327707-25G
77469-428EA 397.58 USD
77469-428 77482-104 77492-508 77502-384
1-(4'-Methyl[1,1'-biphenyl]-4-yl)-1-propanone ≥95%
1-(4'-Methyl[1,1'-biphenyl]-4-yl)-1-propanone

Precaución: For research use only

Formula: C₁₆H₁₆O
MW: 224.3 g/mol
Storage Temperature: Ambient
CAS Number: 96187-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR