1-(5-Chloro-2-fluoro-3-methylphenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

327173-1G 327173-500MG 327173-5G
77469-416EA 415.09 USD
77469-416 77482-092 77492-496
1-(5-Chloro-2-fluoro-3-methylphenyl)butan-1-ol ≥95%
1-(5-Chloro-2-fluoro-3-methylphenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
CAS Number: 3002525-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR