1-(3-Bromo-6-chloro-2-fluorophenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

327164-1G 327164-500MG 327164-5G
77469-412EA 415.09 USD
77469-412 77482-088 77492-492
1-(3-Bromo-6-chloro-2-fluorophenyl)prop-2-en-1-ol ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₉H₇BrClFO
MW: 265.507 g/mol
Storage Temperature: Ambient
CAS Number: 3002422-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR