(3-Bromo-6-chloro-2-fluorophenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

327158-1G 327158-500MG 327158-5G
77469-408EA 415.09 USD
77469-408 77482-084 77492-488
(3-Bromo-6-chloro-2-fluorophenyl)(cyclopropyl)methanol ≥95%
(3-Bromo-6-chloro-2-fluorophenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₀H₉BrClFO
MW: 279.533 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR