(3-Bromo-6-chloro-2-fluorophenyl)(propyl)sulfane ≥96%

Supplier: AOBChem USA

Warning

327156-1G 327156-500MG 327156-5G
77469-404EA 163.77 USD
77469-404 77482-080 77492-484
(3-Bromo-6-chloro-2-fluorophenyl)(propyl)sulfane ≥96%
(3-Bromo-6-chloro-2-fluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₉BrClFS
MW: 283.588 g/mol
Storage Temperature: Ambient
CAS Number: 3002456-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR