(3-(Benzyloxy)-2,6-dichlorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

327145-1G 327145-500MG 327145-5G
77469-400EA 415.09 USD
77469-400 77482-076 77492-480
(3-(Benzyloxy)-2,6-dichlorophenyl)triethylsilane ≥95%
(3-(Benzyloxy)-2,6-dichlorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₉H₂₄Cl₂OSi
MW: 367.385 g/mol
Storage Temperature: Ambient
CAS Number: 3002441-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR