1-(3-(Benzyloxy)-2,6-dichlorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

327139-1G 327139-500MG 327139-5G
77469-396EA 415.09 USD
77469-396 77482-072 77492-476
1-(3-(Benzyloxy)-2,6-dichlorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-(Benzyloxy)-2,6-dichlorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₈H₂₀Cl₂O₂
MW: 339.256 g/mol
Storage Temperature: Ambient
CAS Number: 3002476-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR