(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

327132-1G 327132-500MG 327132-5G
77469-392EA 415.09 USD
77469-392 77482-068 77492-472
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₃BrF₄O
MW: 377.171 g/mol
Storage Temperature: Ambient
CAS Number: 3002498-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR