5'-bromo-2'-fluoro-3'-(trifluoromethyl)-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%

Supplier: AOBChem USA

Warning

327125-1G 327125-500MG 327125-5G
77469-388EA 415.09 USD
77469-388 77482-064 77492-468
5'-bromo-2'-fluoro-3'-(trifluoromethyl)-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%
5'-bromo-2'-fluoro-3'-(trifluoromethyl)-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol

Precaución: For research use only

Formula: C₁₃H₁₁BrF₄O
MW: 339.123 g/mol
Storage Temperature: Ambient
CAS Number: 3002480-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR