(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

327123-1G 327123-500MG 327123-5G
77469-384EA 415.09 USD
77469-384 77482-060 77492-464
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-methoxyphenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₁BrF₄O₂
MW: 379.144 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR