(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

327121-1G 327121-500MG 327121-5G
77469-380EA 415.09 USD
77469-380 77482-056 77492-460
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₉BrF₄O
MW: 313.086 g/mol
Storage Temperature: Ambient
CAS Number: 3002504-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR