1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

327115-1G 327115-500MG 327115-5G
77469-376EA 415.09 USD
77469-376 77482-052 77492-456
1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-one ≥95%
1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-one

Precaución: For research use only

Formula: C₁₀H₇BrF₄O
Storage Temperature: Ambient
CAS Number: 3002422-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR