1-(2,4-Dichlorothiazol-5-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

327092-1G 327092-500MG 327092-5G
77469-372EA 782.8 USD
77469-372 77482-048 77492-452
1-(2,4-Dichlorothiazol-5-yl)ethanone ≥95%
1-(2,4-Dichlorothiazol-5-yl)ethanone

Precaución: For research use only

Formula: C₅H₃Cl₂NOS
MW: 196.054 g/mol
Storage Temperature: Ambient
CAS Number: 1823383-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR