2,4-Dichloro-5-iodothiazole ≥95%

Supplier: AOBChem USA

Warning

327084-1G 327084-500MG 327084-5G
77469-368EA 715.85 USD
77469-368 77482-044 77492-448
2,4-Dichloro-5-iodothiazole ≥95%
2,4-Dichloro-5-iodothiazole

Precaución: For research use only

Formula: C₃Cl₂INS
Storage Temperature: Ambient
CAS Number: 2680537-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR