2-Isopropoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32673-1G 32673-500MG 32673-5G
77469-356EA 200.85 USD
77469-356 77482-032 77492-436
2-Isopropoxy-3-methylbenzaldehyde ≥97%
2-Isopropoxy-3-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD23699166
CAS Number: 532965-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR