1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

325574-1G 325574-500MG 325574-5G
77469-352EA 415.09 USD
77469-352 77482-028 77492-432
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₇F₅O
Storage Temperature: Ambient
CAS Number: 3002498-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR