Cyclopropyl(2,3-difluoro-4-(trifluoromethyl)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

325570-1G 325570-500MG 325570-5G
77469-348EA 415.09 USD
77469-348 77482-024 77492-428
Cyclopropyl(2,3-difluoro-4-(trifluoromethyl)phenyl)methanol ≥95%
Cyclopropyl(2,3-difluoro-4-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₁₁H₉F₅O
MW: 252.181 g/mol
Storage Temperature: Ambient
CAS Number: 3002541-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR