1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

325567-1G 325567-500MG 325567-5G
77469-344EA 415.09 USD
77469-344 77482-020 77492-424
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃F₅O
Storage Temperature: Ambient
CAS Number: 3002441-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR