(2,3-Difluoro-4-(trifluoromethyl)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

325561-1G 325561-500MG 325561-5G
77469-340EA 415.09 USD
77469-340 77482-016 77492-420
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(ethyl)sulfane ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₇F₅S
Storage Temperature: Ambient
CAS Number: 3002465-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR