1-(p-tolyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

325477-1G 325477-500MG 325477-5G
77469-332EA 341.96 USD
77469-332 77482-008 77492-412
1-(p-tolyl)propan-2-ol ≥95%
1-(p-tolyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
CAS Number: 5040-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR