3-Chloro-2-hydroxy-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32529-1G 32529-500MG 32529-5G
77469-328EA 264.71 USD
77469-328 77482-004 77492-408
3-Chloro-2-hydroxy-5-methoxybenzaldehyde ≥97%
3-Chloro-2-hydroxy-5-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD15143231
CAS Number: 90110-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR