2-Chloro-5-fluoro-3-(trifluoromethyl)-benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

32494-1G 32494-500MG 32494-5G
77469-324EA 224.54 USD
77469-324 77482-000 77492-404
2-Chloro-5-fluoro-3-(trifluoromethyl)-benzaldehyde ≥95%
2-Chloro-5-fluoro-3-(trifluoromethyl)-benzaldehyde

Precaución: For research use only

Formula: C₈H₃ClF₄O
MW: 226.555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529812
CAS Number: 95445-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR