2-Ethoxy-6-fluoro-phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxy-6-fluorophenol

32468-1G 32468-500MG 32468-5G
77469-320EA 284.28 USD
77469-320 77481-996 77492-400
2-Ethoxy-6-fluoro-phenol ≥97%
2-Ethoxy-6-fluoro-phenol

Precaución: For research use only

Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16878639
CAS Number: 868761-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR