Methyl 2-fluoro-3,5-diMethyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

324471-1G 324471-500MG 324471-5G
77469-316EA 591.22 USD
77469-316 77481-992 77492-396
Methyl 2-fluoro-3,5-diMethyl-α-oxobenzeneacetate ≥95%
Methyl 2-fluoro-3,5-diMethyl-α-oxobenzeneacetate

Precaución: For research use only

Formula: C₁₁H₁₁FO₃
MW: 210.2 g/mol
Storage Temperature: Ambient
CAS Number: 2091155-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR