(E)-ethyl 4-(4-bromophenyl)-4-oxobut-2-enoate ≥95%

Supplier: AOBChem USA

Warning

324169-1G 324169-500MG 324169-5G
77469-312EA 479.98 USD
77469-312 77481-988 77492-390
(E)-ethyl 4-(4-bromophenyl)-4-oxobut-2-enoate ≥95%
(E)-ethyl 4-(4-bromophenyl)-4-oxobut-2-enoate

Precaución: For research use only

Formula: C₁₂H₁₁BrO₃
MW: 283.1179 g/mol
Storage Temperature: Ambient
CAS Number: 35338-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR