2-Chloro-1-(3-isopropylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

324136-1G 324136-500MG 324136-5G
77469-308EA 509.85 USD
77469-308 77481-984 77492-386
2-Chloro-1-(3-isopropylphenyl)ethanone ≥95%
2-Chloro-1-(3-isopropylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₃ClO
MW: 196.6733 g/mol
Storage Temperature: Ambient
CAS Number: 1824050-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR