1-(4-butoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

323942-1G 323942-500MG 323942-5G
77469-304EA 415.09 USD
77469-304 77481-980 77492-382
1-(4-butoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-butoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Storage Temperature: Ambient
CAS Number: 1980420-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR